2-Morpholinopyrimidin-5-ylboronic acid 95%

Lieferant: Apollo Scientific

OR360280-250MG
APOSOR360280-250MGEA 75.4 EUR
APOSOR360280-250MG
2-Morpholinopyrimidin-5-ylboronic acid 95%
2-Morpholinopyrimidin-5-ylboronic acid
Formel: C₈H₁₂BN₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD07375148
CAS-Nummer: 870521-33-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR