2-Isopropoxy-5-pyridinboronsäurepinakolester 98%

Lieferant: Apollo Scientific

Synonyme: 2-Isopropoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(6-Isopropoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-Isopropoxy-3-pyridylboronic acid Pinacol ester, 2-Isopropoxypyridine-5-boronic acid pinacol ester

OR360292-5G
APOSOR360292-5GEA 212 EUR
APOSOR360292-5G
2-Isopropoxy-5-pyridinboronsäurepinakolester 98%
2-Isopropoxy-5-pyridinboronsäurepinakolester
Formel: C₁₄H₂₂BNO₃
Molecular Weight: 263.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368871
CAS-Nummer: 871839-91-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR