2-(4-Morpholino)-5-pyrimidinboronsäurepinakolester 97%

Lieferant: Apollo Scientific

Synonyme: 4-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl)morpholine, 2-(4-Morpholino)pyrimidine-5-boronic acid, pinacol eser, 2-(4-Morpholino)pyrimidine-5-boronic acid, pinacol ester

OR360308-1G
APOSOR360308-1GEA 62.5 EUR
APOSOR360308-1G
2-(4-Morpholino)-5-pyrimidinboronsäurepinakolester 97%
2-(4-Morpholino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₄H₂₂BN₃O₃
Molecular Weight: 291,1538 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368246
CAS-Nummer: 957198-30-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR