2-Morpholino-5-pyridinboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR360316-1G
OR360316-25G
OR360316-5G
APOSOR360316-1GEA
27.7
EUR
APOSOR360316-1G
APOSOR360316-25G
APOSOR360316-5G
2-Morpholino-5-pyridinboronsäure 98%
2-Morpholino-5-pyridinboronsäure
Formel:
C₉H₁₃BN₂O₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD03086116 CAS-Nummer: 904326-93-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...