2-Morpholino-5-pyridinboronsäure 98%

Lieferant: Apollo Scientific
Achtung

OR360316-1G OR360316-25G OR360316-5G
APOSOR360316-1GEA 27.7 EUR
APOSOR360316-1G APOSOR360316-25G APOSOR360316-5G
2-Morpholino-5-pyridinboronsäure 98%
2-Morpholino-5-pyridinboronsäure
Formel: C₉H₁₃BN₂O₃
Lagertemperatur: Kühlschrank
MDL: MFCD03086116
CAS-Nummer: 904326-93-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR