2-Amino-4-methyl-5-pyrimidinboronsäurepinakolester 95+%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester, 4-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine, 4-Methyl-2-aminopyrimidine-5-boronic acid, pinacol ester

OR360325-250MG
APOSOR360325-250MGEA 40.6 EUR
APOSOR360325-250MG
2-Amino-4-methyl-5-pyrimidinboronsäurepinakolester 95+%
2-Amino-4-methyl-5-pyrimidinboronsäurepinakolester
Formel: C₁₁H₁₈BN₃O₂
Molecular Weight: 235.09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18072554
CAS-Nummer: 944401-55-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR