1-Isobutyl-1H-pyrazole-4-boronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (1-Isobutyl-1H-pyrazol-4-yl)boronic acid

OR360330-1G
APOSOR360330-1GEA 53.8 EUR
APOSOR360330-1G
1-Isobutyl-1H-pyrazole-4-boronic acid 98%
1-Isobutyl-1H-pyrazole-4-boronic acid
Formel: C₇H₁₃BN₂O₂
Molecular Weight: 168,001 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09951916
CAS-Nummer: 929094-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR