2-(Ethoxycarbonyl)furan-5-boronsäure 98%

Lieferant: Apollo Scientific

Synonyme: (5-(Ethoxycarbonyl)furan-2-yl)boronic acid

OR360332-1G
APOSOR360332-1GEA 69.6 EUR
APOSOR360332-1G
2-(Ethoxycarbonyl)furan-5-boronsäure 98%
2-(Ethoxycarbonyl)furan-5-boronsäure
Formel: C₇H₉BO₅
Molecular Weight: 183.9544 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10696632
CAS-Nummer: 1150114-44-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR