1-Phenyl-1H-pyrazol-4-carbaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Phenylpyrazole-4-carboxaldehyde, 1-Phenyl-1H-pyrazole-4-carboxaldehyde
OR3604-1G
APOSOR3604-1GEA
30.6
EUR
APOSOR3604-1G
1-Phenyl-1H-pyrazol-4-carbaldehyd 98%
1-Phenyl-1H-pyrazol-4-carbaldehyd
Formel:
C₁₀H₈N₂O Molecular Weight: 172,18 g/mol Schmelzpunkt: 84…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179568 CAS-Nummer: 54605-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...