2,4,6-Triisopropylphenylboronsäuredimethylester 96%

Lieferant: Apollo Scientific

Synonyme: Dimethyl (2,4,6-triisopropylphenyl)boronate

OR360429-250MG
APOSOR360429-250MGEA 116 EUR
APOSOR360429-250MG
2,4,6-Triisopropylphenylboronsäuredimethylester 96%
2,4,6-Triisopropylphenylboronsäuredimethylester
Formel: C₁₇H₂₉BO₂
Molecular Weight: 276,2220 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09265170
CAS-Nummer: 145434-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR