4-(2-Methylphenyl)phenylboronic acid 95+%

Lieferant: Apollo Scientific

Synonyme: (2'-Methyl-[1,1'-biphenyl]-4-yl)boronic acid

OR360438-250MG
APOSOR360438-250MGEA 48 EUR
APOSOR360438-250MG
4-(2-Methylphenyl)phenylboronic acid 95+%
4-(2-Methylphenyl)phenylboronic acid
Formel: C₁₃H₁₃BO₂
Molecular Weight: 212.052 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08701726
CAS-Nummer: 491595-36-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR