9,10-Bis(2-naphthyl)anthracene-2-ylboronic acid 95+%

Lieferant: Apollo Scientific

Synonyme: (9,10-Di(naphthalen-2-yl)anthracen-2-yl)boronic acid

OR360482-1G
APOSOR360482-1GEA 60.9 EUR
APOSOR360482-1G
9,10-Bis(2-naphthyl)anthracene-2-ylboronic acid 95+%
9,10-Bis(2-naphthyl)anthracene-2-ylboronic acid
Formel: C₃₄H₂₃BO₂
Molecular Weight: 474,356 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11045063
CAS-Nummer: 867044-28-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR