4-Isoxazolboronsäure 98%
Lieferant: Apollo Scientific
Synonyme:
Isoxazol-4-ylboronic acid
OR360562-1G
OR360562-5G
APOSOR360562-1GEA
62.5
EUR
APOSOR360562-1G
APOSOR360562-5G
4-Isoxazolboronsäure 98%
Isoxazole-4-boronic acid
Formel:
C₃H₄BNO₃ Molecular Weight: 112.8798 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06657892 CAS-Nummer: 1008139-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...