(S)-2,2'-Dimethoxy-1,1'-binaphthalin-3,3'-diboronsäure 98%

Lieferant: Apollo Scientific

Synonyme: (2,2'-Dimethoxy-[1,1'-binaphthalene]-3,3'-diyl)diboronic acid, (S)-2,2'-Dimethoxy-1,1'-binaphthyl-3,3'-diboronic acid

OR360578-1G
APOSOR360578-1GEA 144 EUR
APOSOR360578-1G
(S)-2,2'-Dimethoxy-1,1'-binaphthalin-3,3'-diboronsäure 98%
(S)-2,2'-Dimethoxy-1,1'-binaphthalin-3,3'-diboronsäure
Formel: C₂₂H₂₀B₂O₆
Molecular Weight: 402,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027249
CAS-Nummer: 220204-00-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR