3-Allyloxyphenylboronic acid 97%
Lieferant: Apollo Scientific
OR360632-1G
APOSOR360632-1GEA
128
EUR
APOSOR360632-1G
3-Allyloxyphenylboronic acid 97%
3-Allyloxyphenylboronic acid
Formel:
C₉H₁₁BO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11103733 CAS-Nummer: 222840-95-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...