2-Amino-4-chlorophenylboronic acid, pinacol ester 97%

Lieferant: Apollo Scientific

Synonyme: 5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

OR360635-1G
APOSOR360635-1GEA 92.8 EUR
APOSOR360635-1G
2-Amino-4-chlorophenylboronic acid, pinacol ester 97%
2-Amino-4-chlorophenylboronic acid, pinacol ester
Formel: C₁₂H₁₇BClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11617915
CAS-Nummer: 863578-21-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR