4-Amino-1-naphthalinboronsäurepinakolester 97%
Lieferant: Apollo Scientific
Synonyme:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-1-amine
OR360653-250MG
APOSOR360653-250MGEA
116
EUR
APOSOR360653-250MG
4-Amino-1-naphthalinboronsäurepinakolester 97%
4-Amino-1-naphthalinboronsäurepinakolester
Formel:
C₁₆H₂₀BNO₂ Molecular Weight: 269.1465 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15143602 CAS-Nummer: 1218790-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...