4-Amino-3-nitrophenylboronsäurepinakolester 98%

Lieferant: Apollo Scientific

OR360654-5G
APOSOR360654-5GEA 213 EUR
APOSOR360654-5G
4-Amino-3-nitrophenylboronsäurepinakolester 98%
4-Amino-3-nitrophenylboronsäurepinakolester
Formel: C₁₂H₁₇BN₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD06795680
CAS-Nummer: 833486-94-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR