Benzo[b]furan-3-boronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzofuran-3-ylboronic acid

OR360664-5G
APOSOR360664-5GEA 148 EUR
APOSOR360664-5G
Benzo[b]furan-3-boronsäure 98%
Benzo[b]furan-3-boronsäure
Formel: C₈H₇BO₃
Molecular Weight: 161.95 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD06801687
CAS-Nummer: 317830-83-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR