Benzofuran-7-boronic acid, pinacol ester 97%

Lieferant: Apollo Scientific

OR360666-250MG
APOSOR360666-250MGEA 116 EUR
APOSOR360666-250MG
Benzofuran-7-boronic acid, pinacol ester 97%
Benzofuran-7-boronic acid, pinacol ester
Formel: C₁₄H₁₇BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD13810042
CAS-Nummer: 1192755-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR