1-Boc-5-chlor-3-indolboronsäurepinakolester 95+%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 5-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

OR360719-250MG
APOSOR360719-250MGEA 72.5 EUR
APOSOR360719-250MG
1-Boc-5-chlor-3-indolboronsäurepinakolester 95+%
1-Boc-5-chlor-3-indolboronsäurepinakolester
Formel: C₁₉H₂₅BClNO₄
Molecular Weight: 377,6701 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15143612
CAS-Nummer: 1218790-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR