1-Boc-6-chlor-3-indolboronsäurepinakolester 98%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 6-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

OR360720-250MG
APOSOR360720-250MGEA 248 EUR
APOSOR360720-250MG
1-Boc-6-chlor-3-indolboronsäurepinakolester 98%
1-Boc-6-chlor-3-indolboronsäurepinakolester
Formel: C₁₉H₂₅BClNO₄
Molecular Weight: 377,6701 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15143606
CAS-Nummer: 1218790-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR