Boric acid, pinacol ester 97%

Lieferant: Apollo Scientific
Gefahr

OR360754-5G OR360754-1G
APOSOR360754-5GEA 330 EUR
APOSOR360754-5G APOSOR360754-1G
Boric acid, pinacol ester 97%
Boric acid, pinacol ester
Formel: C₆H₁₃BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD15143603
CAS-Nummer: 25240-59-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR