5-Carboxyfuran-2-boronic acid 96%

Lieferant: Apollo Scientific

Synonyme: 5-Boronofuran-2-carboxylic acid

OR360876-250MG
APOSOR360876-250MGEA 48 EUR
APOSOR360876-250MG
5-Carboxyfuran-2-boronic acid 96%
5-Carboxyfuran-2-boronic acid
Formel: C₅H₅BO₅
Molecular Weight: 155.901 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD04113807
CAS-Nummer: 852228-11-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR