2-Carboxyphenylboronic acid, pinacol ester 97%

Lieferant: Apollo Scientific

Synonyme: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

OR360890-5G
APOSOR360890-5GEA 102 EUR
APOSOR360890-5G
2-Carboxyphenylboronic acid, pinacol ester 97%
2-Carboxyphenylboronic acid, pinacol ester
Formel: C₁₃H₁₇BO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD08458199
CAS-Nummer: 1187591-17-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR