2,1,3-Benzoxadiazole-5-carbohydrazide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,1,3-Benzoxadiazole-5-carboxylic acid hydrazide, Benzofurazan-5-carboxylic acid hydrazide
OR3609-1G
APOSOR3609-1GEA
219
EUR
APOSOR3609-1G
2,1,3-Benzoxadiazole-5-carbohydrazide
2,1,3-Benzoxadiazole-5-carbohydrazide
Formel:
C₇H₆N₄O₂ Molecular Weight: 178,148 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00276976 CAS-Nummer: 175203-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...