4-Chloro-2-methylphenylboronic acid, pinacol ester 95+%

Lieferant: Apollo Scientific

OR360972-5G
APOSOR360972-5GEA 55.1 EUR
APOSOR360972-5G
4-Chloro-2-methylphenylboronic acid, pinacol ester 95+%
4-Chloro-2-methylphenylboronic acid, pinacol ester
Formel: C₁₃H₁₈BClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18729895
CAS-Nummer: 1030832-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR