4-(Cyanmethoxy)phenylboronsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-(Cyanomethoxy)phenyl)boronic acid

OR361025-250MG
APOSOR361025-250MGEA 26.1 EUR
APOSOR361025-250MG
4-(Cyanmethoxy)phenylboronsäure 95%
4-(Cyanmethoxy)phenylboronsäure
Formel: C₈H₈BNO₃
Molecular Weight: 176,9700 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09908170
CAS-Nummer: 947533-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR