2,4-Dichlor-5-methoxyphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2,4-Dichloro-5-methoxyphenyl)boronic acid
OR361110-1G
APOSOR361110-1GEA
180
EUR
APOSOR361110-1G
2,4-Dichlor-5-methoxyphenylboronsäure 98%
2,4-Dichlor-5-methoxyphenylboronsäure
Formel:
C₇H₇BCl₂O₃ Molecular Weight: 220.85 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12546454 CAS-Nummer: 431942-67-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...