2,4-Dichlorphenylboronsäurepinakolester 96%

Lieferant: Apollo Scientific

Synonyme: 2-(2,4-Dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR361114-5G
APOSOR361114-5GEA 151 EUR
APOSOR361114-5G
2,4-Dichlorphenylboronsäurepinakolester 96%
2,4-Dichlorphenylboronsäurepinakolester
Formel: C₁₂H₁₅BCl₂O₂
Molecular Weight: 272.97 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06795660
CAS-Nummer: 68716-50-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR