2,3-Dihydro-1-benzofuran-7-boronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (2,3-Dihydrobenzofuran-7-yl)boronic acid

OR361124-1G
APOSOR361124-1GEA 72.5 EUR
APOSOR361124-1G
2,3-Dihydro-1-benzofuran-7-boronic acid 98%
2,3-Dihydro-1-benzofuran-7-boronic acid
Formel: C₈H₉BO₃
Molecular Weight: 163,966 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06801700
CAS-Nummer: 685514-61-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR