2,3-Dihydrobenzofuran-7-boronic acid, pinacol ester 98%

Lieferant: Apollo Scientific

Synonyme: 2-(2,3-Dihydrobenzofuran-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR361126-250MG
APOSOR361126-250MGEA 77 EUR
APOSOR361126-250MG
2,3-Dihydrobenzofuran-7-boronic acid, pinacol ester 98%
2,3-Dihydrobenzofuran-7-boronic acid, pinacol ester
Formel: C₁₄H₁₉BO₃
Molecular Weight: 246,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109314
CAS-Nummer: 934586-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR