2,6-Dimethyl-4-pyridinboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2,6-Dimethylpyridin-4-yl)boronic acid
OR361167-1G
APOSOR361167-1GEA
59.6
EUR
APOSOR361167-1G
2,6-Dimethyl-4-pyridinboronsäure
2,6-Dimethyl-4-pyridineboronic acid
Formel:
C₇H₁₀BNO₂ Molecular Weight: 150.9708 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD11042425 CAS-Nummer: 846548-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...