3-((1,3-Dioxolan-2-yl)methoxy)phenylboronic acid, pinacol ester

Lieferant: Apollo Scientific
Achtung

OR361181-250MG
APOSOR361181-250MGEA 128 EUR
APOSOR361181-250MG
3-((1,3-Dioxolan-2-yl)methoxy)phenylboronic acid, pinacol ester
3-((1,3-Dioxolan-2-yl)methoxy)phenylboronic acid, pinacol ester
Formel: C₁₆H₂₃BO₅
Molecular Weight: 306,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06657708
CAS-Nummer: 850411-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR