2-Hydroxymethylthiophen-4-boronic acid, pinacol ester 97%

Lieferant: Apollo Scientific

Synonyme: (4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl)methanol

OR361268-250MG
APOSOR361268-250MGEA 72.5 EUR
APOSOR361268-250MG
2-Hydroxymethylthiophen-4-boronic acid, pinacol ester 97%
2-Hydroxymethylthiophen-4-boronic acid, pinacol ester
Formel: C₁₁H₁₇BO₃S
Molecular Weight: 240,127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08706011
CAS-Nummer: 864754-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR