6-Indolboronsäurepinakolester 95+%

Lieferant: Apollo Scientific

Synonyme: 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

OR361278-5G
APOSOR361278-5GEA 207 EUR
APOSOR361278-5G
6-Indolboronsäurepinakolester 95+%
6-Indolboronsäurepinakolester
Formel: C₁₄H₁₈BNO₂
Molecular Weight: 243.1092 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368852
CAS-Nummer: 642494-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR