1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester 98%

Lieferant: Apollo Scientific
Achtung

OR361314-5G
APOSOR361314-5GEA 63.8 EUR
APOSOR361314-5G
1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester 98%
1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester
Formel: C₁₂H₂₁BN₂O₂
Schmelzpunkt: 36…44 °C
Lagertemperatur: Kühlschrank
MDL: MFCD11867881
CAS-Nummer: 879487-10-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR