4-(4'-Methoxybenzyloxy)phenylboronsäure 96%

Lieferant: Apollo Scientific

Synonyme: (4-((4-Methoxybenzyl)oxy)phenyl)boronic acid

OR361323-5G
APOSOR361323-5GEA 169 EUR
APOSOR361323-5G
4-(4'-Methoxybenzyloxy)phenylboronsäure 96%
4-(4'-Methoxybenzyloxy)phenylboronsäure
Formel: C₁₄H₁₅BO₄
Molecular Weight: 258,0800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09038417
CAS-Nummer: 156635-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR