4-(Methoxycarbonylmethyl)phenylboronsäure 97%

Lieferant: Apollo Scientific

Synonyme: (4-(2-Methoxy-2-oxoethyl)phenyl)boronic acid

OR361341-1G
APOSOR361341-1GEA 108 EUR
APOSOR361341-1G
4-(Methoxycarbonylmethyl)phenylboronsäure 97%
4-(Methoxycarbonylmethyl)phenylboronsäure
Formel: C₉H₁₁BO₄
Molecular Weight: 193,9922 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09997796
CAS-Nummer: 454185-96-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR