3-Methoxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester 96%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-Methoxy-4-(4,4,5,5-tetraMethyl-1,3,2-dioxaborolan-2-yl)benzoate

OR361370-1G
APOSOR361370-1GEA 72.5 EUR
APOSOR361370-1G
3-Methoxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester 96%
3-Methoxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester
Formel: C₁₅H₂₁BO₅
Molecular Weight: 292.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06797992
CAS-Nummer: 603122-40-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR