2-Methoxy-6-methylpyridine-3-boronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (2-Methoxy-6-methylpyridin-3-yl)boronic acid

OR361377-250MG
APOSOR361377-250MGEA 72.5 EUR
APOSOR361377-250MG
2-Methoxy-6-methylpyridine-3-boronic acid 98%
2-Methoxy-6-methylpyridine-3-boronic acid
Formel: C₇H₁₀BNO₃
Molecular Weight: 166.97 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07374993
CAS-Nummer: 1000802-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR