2-(N-Methylaminomethyl)phenylboronic acid, pinacol ester 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: N-Methyl-1-(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine

OR361411-1G
APOSOR361411-1GEA 94.4 EUR
APOSOR361411-1G
2-(N-Methylaminomethyl)phenylboronic acid, pinacol ester 95%
2-(N-Methylaminomethyl)phenylboronic acid, pinacol ester
Formel: C₁₄H₂₂BNO₂
Molecular Weight: 247,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11855967
CAS-Nummer: 1150271-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR