2-Methylthiazole-5-boronic acid, pinacol ester 97%

Lieferant: Apollo Scientific

OR361488-1G OR361488-250MG
APOSOR361488-1GEA 116 EUR
APOSOR361488-1G APOSOR361488-250MG
2-Methylthiazole-5-boronic acid, pinacol ester 97%
2-Methylthiazole-5-boronic acid, pinacol ester
Formel: C₁₀H₁₆BNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD10697442
CAS-Nummer: 1218791-01-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR