4-(2-Morpholinoethyl)phenylboronic acid, pinacol ester 95+%

Lieferant: Apollo Scientific

Synonyme: 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenethyl)morpholine

OR361502-250MG
APOSOR361502-250MGEA 91.5 EUR
APOSOR361502-250MG
4-(2-Morpholinoethyl)phenylboronic acid, pinacol ester 95+%
4-(2-Morpholinoethyl)phenylboronic acid, pinacol ester
Formel: C₁₈H₂₈BNO₃
Molecular Weight: 317.231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04440855
CAS-Nummer: 364794-81-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR