Oxindol-4-boronsäurepinakolester 95+%

Lieferant: Apollo Scientific

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-2-one

OR361514-250MG
APOSOR361514-250MGEA 88.6 EUR
APOSOR361514-250MG
Oxindol-4-boronsäurepinakolester 95+%
Oxindol-4-boronsäurepinakolester
Formel: C₁₄H₁₈BNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11504971
CAS-Nummer: 1150271-44-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR