(E)-1-Pentenylboronic acid pinacol ester

Lieferant: Apollo Scientific

OR361518-25G OR361518-5G OR361518-1G
APOSOR361518-25GEA 934 EUR
APOSOR361518-25G APOSOR361518-5G APOSOR361518-1G
(E)-1-Pentenylboronic acid pinacol ester
(E)-1-Pentenylboronic acid pinacol ester
Formel: C₁₁H₂₁BO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05663882
CAS-Nummer: 161395-96-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR