1-Phenylsulphonyl-1H-pyrazole-4-boronic acid pinacol ester 97%

Lieferant: Apollo Scientific

Synonyme: 1-(Phenylsulphonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

OR361535-1G
APOSOR361535-1GEA 52.2 EUR
APOSOR361535-1G
1-Phenylsulphonyl-1H-pyrazole-4-boronic acid pinacol ester 97%
1-Phenylsulphonyl-1H-pyrazole-4-boronic acid pinacol ester
Formel: C₁₅H₁₉BN₂O₄S
Molecular Weight: 334,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09037504
CAS-Nummer: 1073372-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR