4-Piperazinylphenylboronic acid, pinacol ester 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperazine

OR361543-1G
APOSOR361543-1GEA 130 EUR
APOSOR361543-1G
4-Piperazinylphenylboronic acid, pinacol ester 95%
4-Piperazinylphenylboronic acid, pinacol ester
Formel: C₁₆H₂₅BN₂O₂
Molecular Weight: 288.193 g/mol
Schmelzpunkt: 135…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06795649
CAS-Nummer: 912369-50-7
UN: 3259
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR