2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene
Lieferant: Apollo Scientific
Synonyme:
Pyrene-2,7-diboronic acid, pinacol ester
OR361570-1G
APOSOR361570-1GEA
179
EUR
APOSOR361570-1G
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene
Formel:
C₂₈H₃₂B₂O₄ Molecular Weight: 454,18 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD15143601 CAS-Nummer: 688756-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...