1-(3-Pyridylmethyl)-1H-pyrazole-4-boronic acid pinacol ester 97%

Lieferant: Apollo Scientific
Achtung

OR361579-250MG
APOSOR361579-250MGEA 21.9 EUR
APOSOR361579-250MG
1-(3-Pyridylmethyl)-1H-pyrazole-4-boronic acid pinacol ester 97%
1-(3-Pyridylmethyl)-1H-pyrazole-4-boronic acid pinacol ester
Formel: C₁₅H₂₀BN₃O₂
Schmelzpunkt: 102…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08706029
CAS-Nummer: 864754-21-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR